Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD16/Fcγ Receptor III, Mouse Monoclonal Antibody

Catalog No. MS1085S1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1085S1 500 μL Concentrated
MS1085R1 1 mL Prediluted
MS1085R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MS1085S1 Supplier Epredia™ Supplier No. MS1085S1

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD16/Fcγ Receptor III, Mouse Monoclonal Antibody.

CD16 (also known low affinity Fc γ receptor III for IgG (FcγRIII) or Leu 11) exists as a polypepetide-anchored from (FCγRIIIA or CD16A) on human natural killer (NK) cells and monocytes/ macrophages and as a glycosylphosphatidylinositol (GPI)-anchored form (FcγRIIIB or CD16B) on neutrophils. CD16B is polymorphic and the two alleles are termed NA1 and NA2. CD16 plays a role in signal transduction, NK cell activation and antibody-dependent cellular cytotoxicity.

Host Species: Mouse

Clone: 2H7

Isotype: IgG2a

Species Reactivity: Human. Others not tested.

Epitope: Extracellular

Immunogen: Recombinant protein encoding the external domain of human CD16

Molecular Weight: 50-65 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD16 Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2H7
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein encoding the external domain of human CD16
Purification Method Tonsil
Quantity 500 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.