Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD16 Monoclonal Antibody (3G8), NovaFluor™ Blue 530, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Supplier: LIFE TECHNOLOGIES H006T03B02A

Description
CD16 Monoclonal antibody specifically detects CD16 in Baboon, Chimpanzee, Cynomolgus Monkey, Human, Non-human Primate, Rhesus Monkey samples. It is validated for Flow Cytometry
Specifications
CD16 | |
Monoclonal | |
4 μL/Test | |
PBS with 0.09% sodium azide; pH 7.2 | |
A3RFZ7, O75015, P08637 | |
Mouse | |
100 Tests | |
Primary | |
Baboon, Chimpanzee, Cynomolgus Monkey, Human, Non-human Primate, Rhesus Monkey | |
Liquid |
Flow Cytometry | |
3G8 | |
NovaFluor Blue 530 | |
FCGR3B | |
4833442P21Rik, CD16, CD16-2, CD16A, CD16a antigen, CD16b, CD32, CD32 receptor 2, CDw32, cytolytic trigger molecule G7, Fc fragment of IgG intermediate affinity IV receptor, Fc fragment of IgG low affinity IIIa receptor, Fc fragment of IgG receptor IIIa, Fc fragment of IgG receptor IIIb, Fc fragment of IgG, low affinity III, receptor for (CD16), Fc fragment of IgG, low affinity IIIa, receptor, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), Fc fragment of IgG, low affinity IIIa, receptor for (CD16), Fc fragment of IgG, low affinity IIIa, receptor for (CD16), Fc fragment of IgG, low affinity III, receptor for (CD16), Fc fragment of IgG, low affinity IIIb, receptor (CD16b), Fc gamma receptor III, Fc gamma receptor IIIa, Fc gamma receptor III-A, Fc gamma receptor IIIb, Fc gamma RIIIa, Fc gammaRIV, Fc receptor, IgG, low affinity III, Fc receptor, IgG, low affinity IV, Fc receptor-like 3, Fcg receptor III, FCG2, FCG3, Fc-gamma receptor III-2 (CD 16), Fc-gamma receptor IIIb (CD 16), Fc-gamma receptor IIIb (CD16), Fc-gamma RII-b, Fc-gamma RII-c, fc-gamma RIII, Fc-gamma RIIIa, Fc-gamma RIII-alpha, fc-gamma RIIIb, fc-gamma RIII-beta, Fc-gamma-RIIb, Fc-gamma-RIIc, FcgammaRIII, FcgammaRIII a.1, FcgammaRIII a.2, FcgammaRIII a.3, FcgammaRIIIA, FcgammaRIV, FCGR2, FCGR2A, FCGR2B, FCGR2C, FCGR3, Fcgr3a, FCGR3B, Fcgr4, FCGRIII, FcgRIV, FcR-10, FcRII-b, FcRII-c, FcRIII, FCRIIIA, FCRIIIb, Fcrl3, IGFR3, IgG Fc gamma receptor III, igG Fc receptor III, IgG Fc receptor III-1, igG Fc receptor III-2, IMD20, immunoglobulin G Fc receptor III, LOC100911825, Low affinity immunoglobulin gamma Fc region receptor III, low affinity immunoglobulin gamma Fc region receptor III-A, Low affinity immunoglobulin gamma Fc region receptor III-B, low affinity immunoglobulin gamma Fc region receptor III-like, Low affinity immunoglobulin gamma Fc region receptor IV, neutrophil-specific antigen NA, RP11-5K23.1, transmembrane receptor FcgammaRIII-X | |
Affinity chromatography | |
RUO | |
2214, 2215, 720006 | |
4° C, store in dark, DO NOT FREEZE! | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction