Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD16-2 Monoclonal Antibody (AT137), FITC
GREENER_CHOICE

Catalog No. PIMA528253
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528253 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528253 Supplier Invitrogen™ Supplier No. MA528253
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Rat anti-mouse CD16-2 antibody, clone AT137, recognizes CD16-2, also known as FcRIV.

FcgRIV, also known as CD16-2, is an intermediate-affinity activating receptor for IgG2a and IgG2b. CD16-2 is the mouse homolog of human FcgRIIIA. CD16-2 is a low-affinity IgE receptor for all allotypes and the ligation of FcgRIV by antigen-IgE immune complexes promotes macrophage-mediated phagocytosis and is involved in lung inflammation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD16-2
Applications Flow Cytometry
Classification Monoclonal
Clone AT137
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with 1% BSA and <0.1% sodium azide
Gene Fcgr4
Gene Accession No. A0A0B4J1G0
Gene Alias 4833442P21Rik; CD16-2; Fc fragment of IgG, low affinity IIIa, receptor for (CD16); Fc fragment of IgG, low affinity III, receptor for (CD16); Fc gammaRIV; Fc receptor, IgG, low affinity IV; Fc receptor-like 3; FcgammaRIV; Fcgr3a; Fcgr4; FcgRIV; Fcrl3; low affinity immunoglobulin gamma Fc region receptor III-A; Low affinity immunoglobulin gamma Fc region receptor IV
Gene Symbols Fcgr4
Host Species Rat
Immunogen Fusion Protein consisting of mouse CD16-2 and rat CD4.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246256
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.