Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD16, Mouse Anti-Human, RB613, Clone: 3G8, BD Horizon™
Mouse Monoclonal Antibody
$148.00 - $370.00
Specifications
Antigen | CD16 |
---|---|
Clone | 3G8 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | RB613 |
Description
CD16 Monoclonal antibody specifically detects CD16 in Human (QC Testing), Rhesus,Cynomolgus,Baboon (Tested in Development) samples. It is validated for Flow CytometrySpecifications
CD16 | |
Flow Cytometry | |
RB613 | |
Mouse | |
Human, Rhesus Monkey, Cynomolgus, Baboon | |
CD16;CD16A;FCGR3A;FcγRIIIA;FcRIIIa;CD16B;FCGR3B;FcγRIIIB;FcRIIIb | |
Human polymorphonuclear leukocytes | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
3G8 | |
Monoclonal | |
Purified | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
2214, 2215 | |
IgG1 κ | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title