Learn More
CD16 Mouse anti-Human, Unconjugated, Clone: 1E12E11, Proteintech
Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 667791IG150UL
Description
CD16 is a 50-70-kDa low affinity Fc receptor found on the surface of natural killer cells, neutrophil polymorphonuclear leukocytes, monocytes and macrophages. CD16 mediates antibody-dependent cellular cytotoxicity (ADCC) and other antibody-dependent responses, such as phagocytosis. CD16 has been identified as Fc receptors FcγRIIIa (CD16a) and FcγRIIIb (CD16b), encoded by two nearly identical genes, FCGR3A and the FCGR3B.
Specifications
CD16 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Pig | |
IgG1 |
Immunohistochemistry, ELISA | |
1E12E11 | |
Unconjugated | |
150 μL | |
2214 | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.