Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD16-2 Recombinant Rabbit Monoclonal Antibody (011)
GREENER_CHOICE

Catalog No. PIMA529708
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529708 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529708 Supplier Invitrogen™ Supplier No. MA529708
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse CD16-2/FCGR4.

FcgRIV, also known as CD16-2, is an intermediate-affinity activating receptor for IgG2a and IgG2b. CD16-2 is the mouse homolog of human FcgRIIIA. CD16-2 is a low-affinity IgE receptor for all allotypes and the ligation of FcgRIV by antigen-IgE immune complexes promotes macrophage-mediated phagocytosis and is involved in lung inflammation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD16-2
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone 011
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Fcgr4
Gene Accession No. A0A0B4J1G0
Gene Alias 4833442P21Rik; CD16-2; Fc fragment of IgG, low affinity IIIa, receptor for (CD16); Fc fragment of IgG, low affinity III, receptor for (CD16); Fc gammaRIV; Fc receptor, IgG, low affinity IV; Fc receptor-like 3; FcgammaRIV; Fcgr3a; Fcgr4; FcgRIV; Fcrl3; low affinity immunoglobulin gamma Fc region receptor III-A; Low affinity immunoglobulin gamma Fc region receptor IV
Gene Symbols Fcgr4
Host Species Rabbit
Immunogen Recombinant Mouse CD16-2/FCGR4 protein (Met1-Gln203).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246256
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.