Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD160 Antibody (023), Alexa Fluor™ 647 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290073AF647
Description
CD160 Monoclonal antibody specifically detects CD160 in Human samples. It is validated for ELISA, ELISASpecifications
CD160 | |
Monoclonal | |
Alexa Fluor 647 | |
BY55FLJ46513, CD160 antigenimmunoglobulin superfamily member, CD160 molecule, Natural killer cell receptor BY55 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD160 (NP_008984.1; Met1-Leu158). | |
0.1 mL | |
Adaptive Immunity, Immunology, Innate Immunity, Signal Transduction | |
11126 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
023 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction