Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD160/By55 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS2526R

Description
CD160 is a receptor showing broad specificity for both classical and non-classical MHC class I molecules. CD160 is expressed at the cell surface as a tightly disulfide-linked multimer. It is involved in cell proliferation, cell surface receptor signaling pathways, cellular defense response, and regulation of immune response.Specifications
CD160/By55 | |
Polyclonal | |
Unconjugated | |
CD160 | |
AU045688; By55; Cd160; CD160 antigen; CD160 antigen, soluble form; CD160 molecule; CD160 transmembrane isoform; CD160-delta Ig; natural killer cell BY55; Natural killer cell receptor BY55; natural killer cell receptor, immunoglobulin superfamily member; NK1; NK28; RGD1563598 | |
Rabbit | |
Protein A | |
RUO | |
11126 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O95971 | |
CD160 | |
KLH conjugated synthetic peptide derived from human CD160. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction