Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD160 Monoclonal Antibody (02)
GREENER_CHOICE

Catalog No. PIMA529849
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529849 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529849 Supplier Thermo Scientific Supplier No. MA529849
Add to Cart
Add to Cart

CD160 Monoclonal Antibody (2)

CD160 Monoclonal Antibody for ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD160
Applications ELISA
Classification Monoclonal
Clone 02
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CD160
Gene Accession No. 0
Gene Alias AU045688; BY55; CD160; CD160 antigen; CD160 antigen, soluble form; CD160 molecule; CD160 transmembrane isoform; CD160-delta Ig; natural killer cell BY55; natural killer cell receptor; natural killer cell receptor BY55; natural killer cell receptor, immunoglobulin superfamily member; NK1; NK28; RGD1563598
Gene Symbols CD160
Host Species Mouse
Immunogen Recombinant Rhesus CD160 protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 696832
Target Species Rhesus Macaque
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.