Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD161 Antibody (008), Alexa Fluor™ 647 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP306112AF647
Description
CD161 Monoclonal antibody specifically detects CD161 in Mouse samples. It is validated for ELISASpecifications
CD161 | |
Monoclonal | |
Alexa Fluor 647 | |
CD161, CD161 antigen, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD161 (P27811; Gln67-His227). | |
0.1 mL | |
Innate Immunity, Signal Transduction | |
3820 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
008 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction