Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD161 Antibody (10/78) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280629
Description
CD161 Monoclonal specifically detects CD161 in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, CyTOF-ready.Specifications
CD161 | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 1:10-1:1000, Immunohistochemistry : 1:10-1:500, Immunoprecipitation : 1:10-1:500, Immunohistochemistry-Frozen : 1:10-1:500, CyTOF-ready | |
CD161, CD161 antigen, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Rat | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), CyTOF | |
28764 | |
Unconjugated | |
PBS | |
KLRB1 | |
Purified splenic NK cells from the LEW rat strain. | |
0.1 mg | |
Innate Immunity, Signal Transduction | |
3820 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction