Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD161 Antibody (10/78), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB9766W
This item is not returnable.
View return policy
Description
CD161 Monoclonal antibody specifically detects CD161 in Rat samples. It is validated for Flow CytometrySpecifications
CD161 | |
Monoclonal | |
DyLight 650 | |
CD161, CD161 antigen, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR | |
Purified splenic NK cells from the LEW rat strain | |
0.1 mL | |
Innate Immunity, Signal Transduction | |
3820 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry | |
10/78. | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction