Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD161 Antibody (14F1F11) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
$206.50 - $499.50
Specifications
Antigen | CD161 |
---|---|
Clone | 14F1F11 |
Concentration | 1.0 mg/ml |
Dilution | Western Blot 1-5 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 5 ug/ml |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP214845SS
![]() |
Novus Biologicals
NBP214845SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP214845
![]() |
Novus Biologicals
NBP214845 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
CD161 Monoclonal specifically detects CD161 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CD161 | |
1.0 mg/ml | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Signal Transduction | |
Q12918 | |
3820 | |
Amino acids 68-225 of human CD161 were used as the immunogen for this antibody. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
14F1F11 | |
Western Blot 1-5 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 5 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD161, CD161 antigen, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR | |
KLRB1 | |
IgG1 κ | |
Protein G purified | |
The antibody has not been tested in species samples other than Human. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title