Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD161 Antibody (14F1F11), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP214845G
Description
CD161 Monoclonal specifically detects CD161 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CD161 | |
Monoclonal | |
DyLight 488 | |
Q12918 | |
KLRB1 | |
Amino acids 68-225 of human CD161 were used as the immunogen for this antibody. | |
0.1 mL | |
Signal Transduction | |
3820 | |
Human | |
Purified |
Western Blot, Immunohistochemistry (Paraffin) | |
14F1F11 | |
Western Blot, Immunohistochemistry-Paraffin | |
CD161, CD161 antigen, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
The antibody has not been tested in species samples other than Human. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction