Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD161 Antibody (B199.2), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065298AF405
Description
CD161 Monoclonal specifically detects CD161 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Western Blot (Negative).Specifications
CD161 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD161, CD161 antigen, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
CD161 (B199.2) | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot | |
B199.2 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Western Blot (Negative) | |
Q12918 | |
KLRB1 | |
NB100-65298 recognises the human CD161 cell surface antigen, expressed by almost all NK cells and with a small subset of CD3+ve T cells. | |
0.1 mL | |
Innate Immunity, Signal Transduction | |
3820 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction