Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD161 Mouse anti-Human, BB700, Clone: HP-3G10, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 748239
Description
CD161 Monoclonal antibody specifically detects CD161 in Human samples. It is validated for Flow Cytometry.
Specifications
CD161 | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
CD161 | |
Refrigerate (2 to 8°C) |
Flow Cytometry | |
HP-3G10 | |
Brilliant Blue 700 (BB700) | |
50 μg | |
Primary | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction