Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD161 Mouse anti-Human, FITC, Clone: DX12, BD

Catalog No. BDB340536 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
50 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB340536 50 Tests
BDB556080 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB340536 Supplier BD Biosciences Supplier No. 340536
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD161, clone DX12, is derived from splenocytes from C3H/HeJ mice immunized with murine fibroblasts stably transfected with hNKR-P1A cDNA in the pBJ expression vector and fused with Sp2/0 myeloma cells.

CD161 recognizes the human NKR-P1A antigen, a disulfide-linked homodimer of a 40-kdalton (kDa) glycoprotein reduced to 26 kDa by treatment with N-glycanase.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD161
Applications Flow Cytometry
Classification Monoclonal
Clone DX12
Conjugate FITC
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Human NKR-P1A Transfected Mouse Fibroblasts
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.