Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD161/NK1.1 Antibody (PK-136), PerCP - Chimeric, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320165PCP
Description
CD161/NK1.1 Monoclonal antibody specifically detects CD161/NK1.1 in Mouse samples. It is validated for Flow CytometrySpecifications
CD161/NK1.1 | |
Monoclonal | |
PerCP | |
CD161, CD161 antigen, CD161c, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR | |
The original antibody was generated by immunizing (C3H X BALB)F1 mice with spleen cells enriched for NK-1 cells and bone marrow cells from CE mice. | |
0.1 mL | |
Innate Immunity, Signal Transduction | |
17059 | |
Store at 4°C in the dark. | |
IgG κ |
Flow Cytometry | |
PK-136 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction