Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD161a Mouse anti-Rat, FITC, Clone: 10/78, BD

Catalog No. BDB555008 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555008 0.5 mg
BDB561781 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB555008 Supplier BD Biosciences Supplier No. 555008
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 10/78 antibody reacts with NKR-P1A, a 60-kDa homodimer expressed on all natural killer (NK) cells and a small subset of T lymphocytes. The 10/78 antibody competes with the previously described 3.2.3 antibody for binding to the antigen. NKR-P1A is a type-II integral membrane protein with an extracellular C-type lectin domain, which is an NK cell-activating receptor specific for tumor target cells. Many rat dendritic cells have been shown to express NKR-P1A, and a subpopulation of these cells has cytotoxic activity. NKR-P1A has also been detected at low levels on peripheral blood monocytes, and its expression is upregulated in IFN-γ-activated monocytes, specifically in a subpopulation of large monocytes with phagocytic capacity. Furthermore, activated peripheral blood neutrophils may express a low level of NKR-P1A. In the mouse and rat, three members of the NKR-P1 gene family have been identified; but in the human gene family, a single NKR-P1 homologue has been discovered and designated Cd161

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD161a
Applications Flow Cytometry
Classification Monoclonal
Clone Oct-78
Concentration 0.5mg/mL
Conjugate FITC
Description NKR-P1A
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Not reported
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.