Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD161a Mouse, anti-Rat, R718, Clone: 10/78, BD OptiBuild™

Catalog No. BDB752285 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752285 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752285 Supplier BD Biosciences Supplier No. 752285
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 10/78 monoclonal antibody recognizes the rat CD161 proteins, CD161a (also known as, Klrb1a, or Nkrp1a/NKR-P1A), and CD161b (Klrb1b, Nkrp1b/NKR-P1B). These type II transmembrane glycoproteins have an extracellular C-type lectin domain and thus belong to the C-type lectin superfamily. These CD161 proteins form ∼ 60 kDa homodimers that are expressed on natural killer cells and subsets of T lymphocytes, activated monocytes, and dendritic cells. The 10/78 antibody competes with the previously-described 3.2.3 antibody for binding to these CD161 proteins. CD161 molecules are C-type lectin-like receptors that can either activate (CD161a) or inhibit (CD161b) effector leucocyte responses, eg, cytotoxicity or cytokine production, against target cells which express C-type lectin-like related (Clr) molecules. Although several members of the Klrb1 gene family have been identified in the mouse and rat, only a single human KLRB1 homolog has been discovered.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD161a
Applications Flow Cytometry
Classification Monoclonal
Clone 10/78
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.