Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD162 Mouse anti-Human, BUV395, Clone: KPL-1, BD Optibuild™

Catalog No. BDB743484 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743484 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743484 Supplier BD Biosciences Supplier No. 743484
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The KPL-1 monoclonal antibody specifically binds to p-selectin glycoprotein ligand-1 (PSGL-1), a 120-220 kDa, mucin-like type I transmembrane glycoprotein. CD162 is expressed on neutrophils, monocytes, and most lymphocytes. CD162 binds to CD62P (P-Selectin), CD62E (E-Selectin) and CD62L (L-Selectin). This molecule mediates rolling of leukocytes on activated endothelium and the interaction between leukocytes and activated platelets or other leukocytes found at inflammatory sites. It has also been reported that this clone completely inhibits the interaction between P-Selectin and either CD4-T cells or neutrophils.

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD162
Classification Monoclonal
Clone KPL-1
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 395
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias SELPLG, Selectin P ligand, P-selectin glycoprotein ligand 1, PSGL-1, CLA
Host Species Mouse
Immunogen Human PSGL-1 Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.