Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD162 (PSGL-1) Monoclonal Antibody (4RA10), PerCP-eFluor™ 710, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
$216.00 - $483.00
Specifications
| Antigen | CD162 (PSGL-1) |
|---|---|
| Clone | 4RA10 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
46-162-180
|
Invitrogen™
46162180 |
25 μg |
Each for $216.00
|
|
|||||
46-162-182
![]()
|
Invitrogen™
46162182 |
100 μg |
Each for $483.00
|
|
|||||
Description
CD162 (PSGL-1) is a 120 kDa homodimer protein found on the surface of leukocytes and platelets. CD162 is known to bind CD62P (P-selectin), CD62L (L-selectin), and to a lower degree CD62E (E-selectin). These interactions play a critical role in leukocyte adhesion, rolling and extravasation. CD162 has been shown to be an important immune marker of T cell exhaustion in chronic viral infections and cancer.Specifications
| CD162 (PSGL-1) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| CD162; CLA; Cutaneous lymphocyte associated antigen; cutaneous lymphocyte-associated associated antigen; leukocyte cell surface adhesion molecule; P-selectin glycoprotein ligand 1; P-selectin glycoprotein ligand 1 propeptide; P-selectin glycoprotein ligand-1; Psgl1; Psgl-1; Selectin P ligand; selectin, platelet (p-selectin) ligand; Selp1; SELPG; Selpl; SELPLG | |
| SELPLG | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| 4RA10 | |
| Flow Cytometry | |
| PerCP-eFluor 710 | |
| Rat | |
| Mouse | |
| Q62170 | |
| 20345 | |
| Mouse PSGL-1 human IgG1 fusion protein. | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| SELPLG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title