Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD162 Rat anti-Mouse, BUV615, Clone: 2PH1, BD OptiBuild™

Catalog No. BDB752333 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 x 50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752333 1 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752333 Supplier BD Biosciences Supplier No. 752333
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD162 Monoclonal antibody specifically detects CD162 in Mouse samples. It is validated for Flow Cytometry.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD162
Applications Flow Cytometry
Classification Monoclonal
Clone 2PH1
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 615
Description Ms CD162 BUV615 2PH1 50 μg
Host Species Rat
Immunogen Ovalbumin-conjugated peptide covering amino acids 42 to 60 of mouse PSGL-1
Quantity 1 x 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity CD162
Target Species Mouse
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG1 κ
Reactivity Mouse
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.