Learn More
Invitrogen™ CD162 Recombinant Rabbit Monoclonal Antibody (031), APC

Description
Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Human CD162 (PSGL-1) (Met1-Gly295) and conjugated with APC under optimum conditions, the unreacted APC was removed. Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze! Sodium azide is toxic to cells and should be disposed of properly. Flush with large volumes of water during disposal.
Specifications
Specifications
| Antigen | CD162 |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Clone | 031 |
| Concentration | 100 μg/mL |
| Conjugate | APC |
| Formulation | PBS with 0.5% BSA and 0.09% sodium azide |
| Gene | SELPLG |
| Gene Accession No. | Q14242 |
| Gene Alias | CD162; CLA; Cutaneous lymphocyte associated antigen; cutaneous lymphocyte-associated associated antigen; leukocyte cell surface adhesion molecule; P-selectin glycoprotein ligand 1; P-selectin glycoprotein ligand 1 propeptide; P-selectin glycoprotein ligand-1; Psgl1; Psgl-1; Selectin P ligand; selectin, platelet (p-selectin) ligand; Selp1; SELPG; Selpl; SELPLG |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.