Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD162 Recombinant Rabbit Monoclonal Antibody (8C3X6)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551508
Description
CD162 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
CD162 (PSGL-1) is a 120 kDa homodimer protein found on the surface of leukocytes and platelets. CD162 is known to bind CD62P (P-selectin), CD62L (L-selectin), and to a lower degree CD62E (E-selectin). These interactions play a critical role in leukocyte adhesion, rolling and extravasation. CD162 has been shown to be an important immune marker of T cell exhaustion in chronic viral infections and cancer.
Specifications
CD162 | |
Recombinant Monoclonal | |
0.36 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q14242, Q62170 | |
SELPLG | |
Recombinant protein of human CD162/PSGL-1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
8C3X6 | |
Unconjugated | |
SELPLG | |
CD162; CLA; Cutaneous lymphocyte associated antigen; cutaneous lymphocyte-associated associated antigen; leukocyte cell surface adhesion molecule; P-selectin glycoprotein ligand 1; P-selectin glycoprotein ligand 1 propeptide; P-selectin glycoprotein ligand-1; Psgl1; Psgl-1; Selectin P ligand; selectin, platelet (p-selectin) ligand; Selp1; SELPG; Selpl; SELPLG | |
Rabbit | |
Affinity chromatography | |
RUO | |
20345, 363930, 6404 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction