Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD163 Antibody (5C6-FAT), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals BM4041MFV610
Description
CD163 Monoclonal antibody specifically detects CD163 in Human samples. It is validated for Flow CytometrySpecifications
CD163 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
5C6-FAT | |
Flow Cytometry | |
CD_antigen: CD163, CD163, CD163 molecule, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163 | |
Human CD163. | |
0.1 mL | |
Angiogenesis, Immune System Diseases, Immunology, Inflammation, Innate Immunity | |
9332 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction