Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD163 Antibody (6D3.2F3) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280632
Description
CD163 Monoclonal specifically detects CD163 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
CD163 | |
Monoclonal | |
1 mg/mL | |
Western Blot 2 μg/mL, Flow Cytometry, Immunohistochemistry : 1:200 - 1:500, Immunohistochemistry-Paraffin : 1:200 - 1:500, CyTOF-ready | |
CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Immunogen's sequence similarity with other species: Bovine (88%), Porcine (87%), Canine (87%), Rat (80%) | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2b κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
6D3.2F3 | |
Unconjugated | |
PBS | |
CD163 | |
A synthetic peptide made to the human CD163 protein sequence (between residues 200-400). [Swiss-Prot: Q86VB7] | |
0.1 mg | |
Immune System Diseases, Immunology, Innate Immunity | |
9332 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction