Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD163 Antibody (EDHu-1), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11040686AF700
Description
CD163 Monoclonal specifically detects CD163 in Human, Rat, Porcine, Bovine, Guinea Pig, Primate, Monkey, Rhesus Macaque, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| CD163 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| NB110-40686 recognizes the human CD163 cell surface antigen, a 130-140kD glycoprotein expressed by tissue macrophages. CD163 is not expressed by resting peripheral blood leucocytes, but expression may be induced on monocytes by culture in dexamethasone. | |
| Human, Pig, Bovine, Guinea Pig, Primate, Monkey, Rhesus Macaque, Sheep | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| EDHu-1 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| Q86VB7 | |
| CD163 | |
| Leucocytes harvested from pleural cavity of patients with idiopathic spontaneous pneumothorax. | |
| 0.1 mL | |
| Immune System Diseases, Immunology, Innate Immunity | |
| 9332 | |
| Protein A purified | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction