Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD163 Antibody (EDHu-1) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7199360 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017969 0.025 mg
NB11040686 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017969 Supplier Novus Biologicals Supplier No. NB110406860.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 9 publications

CD163 Monoclonal specifically detects CD163 in Human, Rat, Porcine, Bovine, Guinea Pig, Primate, Monkey, Rhesus Macaque, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay, Flow (Cell Surface).

Specifications

Antigen CD163
Applications Immunofluorescence, Flow Cytometry, Immunohistochemistry, ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone EDHu-1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay, Flow (Cell Surface) 1:10-1:1000
Formulation PBS with 0.09% Sodium Azide
Gene Accession No. Q86VB7
Gene Alias CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130
Gene Symbols CD163
Host Species Mouse
Immunogen Leucocytes harvested from pleural cavity of patients with idiopathic spontaneous pneumothorax.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9332
Test Specificity NB110-40686 recognizes the human CD163 cell surface antigen, a 130-140kD glycoprotein expressed by tissue macrophages. CD163 is not expressed by resting peripheral blood leucocytes, but expression may be induced on monocytes by culture in dexamethasone.
Target Species Sheep, Monkey, Primate, Rhesus Macaque, Guinea Pig, Human, Bovine, Pig
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.