Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD163 Antibody (EDHu-1), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11040686PEATT594
Description
CD163 Monoclonal specifically detects CD163 in Human, Rat, Porcine, Bovine, Guinea Pig, Monkey, Primate, Rhesus Macaque, Sheep samples. It is validated for Flow Cytometry.Specifications
CD163 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
CD163 | |
Leucocytes harvested from pleural cavity of patients with idiopathic spontaneous pneumothorax. | |
0.1 mL | |
Primary | |
NB110-40686 recognizes the human CD163 cell surface antigen, a 130-140kD glycoprotein expressed by tissue macrophages. CD163 is not expressed by resting peripheral blood leucocytes, but expression may be induced on monocytes by culture in dexamethasone. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
EDHu-1 | |
Flow Cytometry | |
CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130 | |
Mouse | |
Protein A purified | |
Immune System Diseases, Immunology, Innate Immunity | |
9332.0 | |
Human, Pig, Bovine, Guinea Pig, Monkey, Primate, Rhesus Macaque, Sheep | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction