Learn More
CD163 Antibody (GHI/61), DyLight 755, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP143341IR
Catalog No. NBP143341IR
Only null left
Add to Cart
Description
Description
CD163 Monoclonal specifically detects CD163 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications
Specifications
CD163
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
CD163
The immunogen for this antibody was CD163.
0.1 mL
Immune System Diseases, Immunology, Innate Immunity
9332
Store at 4C in the dark.
IgG1 κ
Western Blot, Flow Cytometry
GHI/61
Western Blot, Flow Cytometry
CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.