Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD163 Antibody (LND68A), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP260845
Description
CD163 Monoclonal specifically detects CD163 in Bovine, Water Buffalo samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CD163 | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 1:10 - 1:1000 | |
CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130 | |
Mouse | |
Unpurified | |
Immune System Diseases, Immunology, Innate Immunity | |
9332 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Flow Cytometry, Immunohistochemistry, Functional Assay | |
LND68A | |
Unconjugated | |
Ascites with 0.09% Sodium Azide | |
CD163 | |
Bovine lymph node cells | |
0.1 mg | |
Primary | |
Bovine, Water Buffalo | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction