Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD163 Antibody (OTI2B12), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271420H
Description
CD163 Monoclonal specifically detects CD163 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CD163 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
CD163 | |
Full length recombinant protein of human CD163 (NP_004235) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI2B12 | |
Western Blot, Immunohistochemistry | |
CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130 | |
Mouse | |
Affinity Purified | |
Immune System Diseases, Immunology, Innate Immunity | |
9332.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction