Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD163 Antibody (OTI2B12), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP246115
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry, Immunofluorescence
CD163 Monoclonal specifically detects CD163 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.Specifications
CD163 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:200 - 1:4000, Immunohistochemistry 1:150, Immunofluorescence 1:100 | |
CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130 | |
Mouse | |
125.3 kDa | |
0.1 mL | |
Immune System Diseases, Immunology, Innate Immunity | |
9332 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2a |
Western Blot, Immunohistochemistry, Immunofluorescence | |
OTI2B12 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
CD163 | |
Full length human recombinant protein of human CD163(NP_004235) produced in HEK293T cell. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction