Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD163 Antibody (R20) [DyLight 680], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB16072Y
Description
CD163 Monoclonal antibody specifically detects CD163 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
CD163 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
R20 | |
Flow Cytometry, CyTOF-ready | |
CD_antigen: CD163, CD163, CD163 molecule, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163 | |
Balb/c mice immunized with Synovial cells from Rheumatoid Arthritis patients | |
0.1 mL | |
Angiogenesis, Immune System Diseases, Immunology, Inflammation, Innate Immunity | |
9332 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction