Learn More
CD163 Mouse anti-Human, Alexa Fluor™ 488, Clone: MAC2-158 (also known as Mac 2-158; Mac-2–158), BD Pharmingen™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | CD163 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MAC2-158 (also known as Mac 2-158; Mac-2–158) |
| Conjugate | Alexa Fluor 488 |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | CD163; HbSR; M130;MM130;SCARI1;SR-I1 |
| Host Species | Mouse |
| Immunogen | Human Monocytes |
| Purification Method | Affinity Chromatography |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.