Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD163 Antibody (6D3.2F3), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP236495AF647
Description
CD163 Monoclonal specifically detects CD163 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CD163 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CD163 | |
| A synthetic peptide made to the human CD163 protein sequence (between residues 200-400). [Swiss-Prot: Q86VB7] | |
| 0.1 mL | |
| Immune System Diseases, Immunology, Innate Immunity | |
| This antibody is specific for the extracellular domain of human CD163 protein | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 6D3.2F3 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction