Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD163 Mouse anti-Human, Clone: GHI/61, Abnova™

Catalog No. 89933747 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-747 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-747 Supplier Abnova Corporation Supplier No. MAB12601
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against human CD163.

Specifications

Antigen CD163
Applications Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone GHI/61
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human CD163.
Dilution Flow Cytometry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
Formulation In aqueous buffer (Stabilizer, 0.09% sodium azide).
Gene Alias M130/MM130
Gene Symbols CD163
Host Species Mouse
Immunogen Spleen cells from a case of hairy cell leukemia.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9332
Target Species Human
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.