Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD163 Rabbit anti-Human, Unconjugated, Clone: RM371, Abnova™

Catalog No. 89339220 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-220 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-220 Supplier Abnova Corporation Supplier No. MAB22003
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

CD163 Monoclonal antibody specifically detects CD163 in Human samples. It is validated for WB-Ti, IHC-P.

Specifications

Antigen CD163
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM371
Conjugate Unconjugated
Description CD163 molecule
Gene CD163
Gene Alias M130, MM130
Host Species Rabbit
Immunogen A peptide corresponding to N-terminus of human CD163
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 9332
Target Species Human
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.