Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD163L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116626
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116626 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116626 Supplier Invitrogen™ Supplier No. PA5116626
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Rat heart, Rat lung Immunogen sequence: DCLHQNDVSV ICSDGADLEL RLADGSNNCS GRVEVRIHEQ WWTICDQNWK NEQALVVCKQ LGCPFSVFGS RRAKPSNEAR DIWINSISCT GNESALWDCT YDGKAKRTCF RRSDAGVICS DKADLDLRLV GAHSPCYGRL EVKYQGEWGT VCHDRWSTRN AAVVCKQLGC GKPMHVFGMT YFKEASGPIW LDDVSCIGNE S.

This gene encodes a member of the scavenger receptor cysteine-rich (SRCR) superfamily. Members of this family are secreted or membrane-anchored proteins mainly found in cells associated with the immune system. The SRCR family is defined by a 100-110 amino acid SRCR domain, which may mediate protein-protein interaction and ligand binding. The encoded protein contains twelve SRCR domains, a transmembrane region and a cytoplasmic domain. Alternatively spliced transcript variants encoding different isoforms have been described but their full-length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD163L1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CD163L1
Gene Accession No. Q9NR16
Gene Alias CD163 antigen B; CD163 antigen-like 1; CD163 molecule like 1; CD163 molecule-like 1; CD163B; CD163L1; M160; SCARI2; scavenger receptor cysteine-rich type 1 protein M160; UNQ6434/PRO23202
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 350-550 of human CD163L1 (NP_7776012).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283316
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.