Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD164 Antibody (502021) [DyLight 405], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5790E
Description
CD164 Monoclonal antibody specifically detects CD164 in Human samples. It is validated for Flow CytometrySpecifications
CD164 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
502021 | |
Flow Cytometry | |
CD164 antigen, CD164 molecule, sialomucin, Endolyn, MGC-24v, Multi-glycosylated core protein 24, sialomucin | |
NS0 mouse myeloma cell line transfected with human CD14, Asp24-Leu197, Accession # Q04900 | |
0.1 mL | |
Cancer | |
8763 | |
Store at 4°C in the dark. | |
IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction