Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD164 Monoclonal Antibody (05)
GREENER_CHOICE

Catalog No. PIMA529093
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529093 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529093 Supplier Invitrogen™ Supplier No. MA529093
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human CD164.

This gene encodes a transmembrane sialomucin and cell adhesion molecule that regulates the proliferation, adhesion and migration of hematopoietic progenitor cells. The encoded protein also interacts with the C-X-C chemokine receptor type 4 and may regulate muscle development. Elevated expression of this gene has been observed in human patients with Sezary syndrome, a type of blood cancer, and a mutation in this gene may be associated with impaired hearing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD164
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 05
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CD164
Gene Accession No. Q04900
Gene Alias A115; A24; AA589639; AW540279; Cd164; CD164 antigen; CD164 antigen, sialomucin; CD164 molecule; CD164 molecule, sialomucin; CD164 variant C; c-Myc binding protein; deafness, autosomal dominant 66; DFNA66; endolyn; MGC-24; MGC-24v; MSSP; MUC-24; Multi-glycosylated core protein 24; sialomucin core protein 24
Gene Symbols CD164
Host Species Mouse
Immunogen Recombinant Human CD164 Protein (Met1-Asp162).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8763
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.