Learn More
Invitrogen™ CD164 Monoclonal Antibody (502021)

Description
In direct ELISAs, no cross-reactivity with recombinant mouse CD164 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | CD164 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 502021 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 5% trehalose and No Preservative |
| Gene | CD164 |
| Gene Accession No. | Q04900 |
| Gene Alias | A115; A24; AA589639; AW540279; Cd164; CD164 antigen; CD164 antigen, sialomucin; CD164 molecule; CD164 molecule, sialomucin; CD164 variant C; c-Myc binding protein; deafness, autosomal dominant 66; DFNA66; endolyn; MGC-24; MGC-24v; MSSP; MUC-24; Multi-glycosylated core protein 24; sialomucin core protein 24 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.