Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD164 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119029
Change view
Click to view available options
Quantity:
20 μL
200 μL
Catalog No. Quantity
PIPA5119029 200 μL
PIPA5122133 20 μL
2 options
This item is not returnable. View return policy

Catalog No. PIPA5119029

Supplier: Invitrogen™ PA5119029

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CD164 Polyclonal Antibody for Western Blot

This gene encodes a transmembrane sialomucin and cell adhesion molecule that regulates the proliferation, adhesion and migration of hematopoietic progenitor cells. The encoded protein also interacts with the C-X-C chemokine receptor type 4 and may regulate muscle development. Elevated expression of this gene has been observed in human patients with Sezary syndrome, a type of blood cancer, and a mutation in this gene may be associated with impaired hearing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD164
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene CD164
Gene Accession No. Q9QX82
Gene Alias A115; A24; AA589639; AW540279; Cd164; CD164 antigen; CD164 antigen, sialomucin; CD164 molecule; CD164 molecule, sialomucin; CD164 variant C; c-Myc binding protein; deafness, autosomal dominant 66; DFNA66; endolyn; MGC-24; MGC-24v; MSSP; MUC-24; Multi-glycosylated core protein 24; sialomucin core protein 24
Gene Symbols CD164
Host Species Rabbit
Immunogen Recombinant protein Cluster Of Differentiation 164. The antigen corresponds to amino acid range 24-160 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83689
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.