Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD164 Recombinant Rabbit Monoclonal Antibody (006), PE
GREENER_CHOICE

Catalog No. PIMA540926
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA540926 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA540926 Supplier Invitrogen™ Supplier No. MA540926
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

May be stored for twelve months without detectable loss of activity. Protect from prolonged exposure to light and do not freeze. Sodium azide is toxic to cells and should be disposed of properly so flush with large volumes of water during disposal.

This gene encodes a transmembrane sialomucin and cell adhesion molecule that regulates the proliferation, adhesion and migration of hematopoietic progenitor cells. The encoded protein also interacts with the C-X-C chemokine receptor type 4 and may regulate muscle development. Elevated expression of this gene has been observed in human patients with Sezary syndrome, a type of blood cancer, and a mutation in this gene may be associated with impaired hearing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD164
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone 006
Concentration 0.1 mg/mL
Conjugate PE
Formulation PBS with 0.5% BSA and 0.09% sodium azide
Gene CD164
Gene Accession No. Q04900
Gene Alias A115; A24; AA589639; AW540279; Cd164; CD164 antigen; CD164 antigen, sialomucin; CD164 molecule; CD164 molecule, sialomucin; CD164 variant C; c-Myc binding protein; deafness, autosomal dominant 66; DFNA66; endolyn; MGC-24; MGC-24v; MSSP; MUC-24; Multi-glycosylated core protein 24; sialomucin core protein 24
Gene Symbols CD164
Host Species Rabbit
Immunogen Recombinant Human CD164 Protein.
Purification Method Protein A
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8763
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.