Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD164L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567679
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567679 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567679 Supplier Invitrogen™ Supplier No. PA567679
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CD164L2 Polyclonal Antibody for Western Blot

CD164L2 gene ontology (GO) annotation includes cytoplasmic vesicle and cytoplasmic vesicle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD164L2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CD164L2
Gene Accession No. Q6UWJ8
Gene Alias CD164 molecule like 2; CD164 sialomucin-like 2 protein; CD164L2; EAPG6122; LOC100359977; sialomucin core protein 24-like; UNQ6122; UNQ6122/PRO20044
Gene Symbols CD164L2
Host Species Rabbit
Immunogen Synthesized peptide derived from human CD164L2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388611
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.