Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD165 Mouse, anti-Human, R718, Clone: SN2, BD OptiBuild™

Catalog No. BDB752127 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752127 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752127 Supplier BD Biosciences Supplier No. 752127
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The SN2 monoclonal antibody specifically recognizes a 37-42 kDa membrane glycoprotein expressed on a subset of peripheral lymphocytes, monocytes, immature thymocytes and on most platelets. CD165, also known as gp37 or AD2, is expressed at low levels on most thymocytes and thymic epithelial cells. CD165 has been detected on islet cells of the pancreas, Bowman's capsule of the kidney and on central nervous system neurons. SN2 antigen has been reported to be identical, or very similar, to the T-cell acute lymphoblastic leukemia antigen, TALLA-1. CD165 has been reported to play a role in the adhesion between thymocytes and thymic epithelial cells during development.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD165
Applications Flow Cytometry
Classification Monoclonal
Clone SN2
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.