Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD166 (ALCAM) Monoclonal Antibody (3A6), Super BrighT&Trade; 600, eBioscience™
GREENER_CHOICE

Catalog No. 63166842
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-166-842 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 63-166-842 Supplier Invitrogen™ Supplier No. 63166842
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Description: This 3A6 monoclonal antibody reacts with human CD166, which is also known as activated leukocyte cell adhesion molecule (ALCAM). CD166 is an immunoglobulin superfamily cell adhesion molecule that is broadly expressed on developing and adult hematopoietic (e.g., activated T and B cells, monocytes, and dendritic cells) and non-hematopoietic (e.g., neural, endothelial, and epithelial) tissues. CD166 has also been detected in melanoma, colorectal carcinoma, as well as cancers of the breast, prostate, and bladder. CD166 mediates cell-cell adhesion by forming low-affinity homophilic interactions (ALCAM-ALCAM) or high-affinity heterophilic interactions with its ligand CD6 (ALCAM-CD6). CD166 plays a role in development, inflammation, and neutrophil migration. Moreover, its expression can be used as a prognostic marker for several cancers and has been reported to be a cancer stem cell marker. The 3A6 antibody has been reported to induce the internalization of CD166 and its recycling back to the cell surface. Applications Reported: This 3A6 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 3A6 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5...

This gene encodes activated leukocyte cell adhesion molecule (ALCAM), also known as CD166 (cluster of differentiation 166), which is a member of a subfamily of immunoglobulin receptors with five immunoglobulin-like domains (VVC2C2C2) in the extracellular domain. This protein binds to T-cell differentiation antigene CD6, and is implicated in the processes of cell adhesion and migration. Multiple alternatively spliced transcript variants encoding different isoforms have been found.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD166 (ALCAM)
Applications Flow Cytometry
Classification Monoclonal
Clone 3A6
Concentration 5 μL/Test
Conjugate Super Bright 600
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene ALCAM
Gene Accession No. Q13740
Gene Alias activated leukocyte cell adhesion molecule; AI853494; ALCAM; BEN; CD166; CD166 antigen; cell adhesion molecule; DM-GRASP; FLJ38514; HB2; KG-CAM; MEMD; MGC71733; MuSC; Protein DM-GRASP; Protein MEMD; SB-10 antigen; SC1; unnamed protein product
Gene Symbols ALCAM
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 214
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.