Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD166 (ALCAM) Mouse anti-Human, MilliporeSigma™

Catalog No. MABN178525 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN1785-25 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1785-25 Supplier MilliporeSigma Supplier No. MABN178525UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CD166 (ALCAM) Antibody, clone TAG-1A3 is a highly specific mouse monoclonal antibody that targets CD166 and has been tested in Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD166 (ALCAM)
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols ALCAM,MEMD
Host Species Mouse
Immunogen Corneal endothelial cells (hCENC) isolated from donated human cadaves and immunized with monophosphoryl-lipid A trehalose dicorynomycolate (MPLCTDM) adjuvant.
Purification Method Protein G Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone TAG-1A3 specifically detects CD133 in human.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.