Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD166 (ALCAM) Recombinant Rabbit Monoclonal Antibody (6Z4Y2)
GREENER_CHOICE

Catalog No. PIMA538206
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538206 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538206 Supplier Thermo Scientific Supplier No. MA538206
Add to Cart
Add to Cart

CD166 (ALCAM) Recombinant Monoclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD166 (ALCAM)
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 6Z4Y2
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ALCAM
Gene Accession No. O35112, Q13740, Q61490
Gene Alias activated leukocyte cell adhesion molecule; AI853494; ALCAM; BEN; CD166; CD166 antigen; cell adhesion molecule; DM-GRASP; FLJ38514; HB2; KG-CAM; MEMD; MGC71733; MuSC; Protein DM-GRASP; Protein MEMD; SB-10 antigen; SC1; unnamed protein product
Gene Symbols ALCAM
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 484-583 of human CD166/ALCAM (Q13740).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11658, 214, 79559
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.